Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003103.2 | NP_001003103.1 | 1 | 89 | 85 | |
XM_005625450.3 | XP_005625507.1 | 1 | 232 | 85 | ||
XM_022423983.1 | XP_022279691.1 | 1 | 233 | 85 | ||
XM_022423984.1 | XP_022279692.1 | 84 | 85 | |||
GenBank mRNA | AB031276.1 | - | 1 | 83 | 85 | |
AF100705.1 | - | 1 | 85 | |||
AF322416.2 | - | 1 | 89 | 85 | ||
AF322417.2 | - | 89 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |