Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625450.3 | XP_005625507.1 | 1 | 970 | 150 | |
XM_005625451.3 | XP_005625508.1 | 1 | 853 | 150 | ||
XM_022423982.1 | XP_022279690.1 | 1 | 827 | 150 | ||
XM_022423983.1 | XP_022279691.1 | 2 | 806 | 150 | ||
XM_022423984.1 | XP_022279692.1 | 1 | 909 | 150 | ||
GenBank mRNA | AB031276.1 | - | 1 | 821 | 150 | |
AF100705.1 | - | 1 | 827 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |