Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRSP-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002947.2 | NP_001002947.1 | 1 - 2 | 106 | 87 | |
GenBank mRNA | AB125103.1 | - | 1 - 2 | 109 | 87 | |
AB294189.1 | - | 1 - 2 | 140 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |