Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003968.1 | NP_001003968.1 | 2 - 3 | 199 | 71 | |
XM_022424909.1 | XP_022280617.1 | 3 - 4 | 452 | 71 | ||
GenBank mRNA | AB162850.1 | - | 2 - 3 | 199 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |