Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010949.1 | NP_001010949.1 | 1 - 2 | 60 | 92 | |
GenBank mRNA | AB183191.1 | - | 1 - 2 | 60 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Inventoried Probe spans exons |