Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003183.1 | NP_001003183.1 | 3 - 4 | 399 | 86 | |
GenBank mRNA | AB191461.1 | - | 3 - 4 | 429 | 86 | |
AF045773.1 | - | 3 - 4 | 399 | 86 | ||
U96127.1 | - | 3 - 4 | 344 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |