Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006953.2 | NP_001006954.1 | 1 | 233 | 107 | |
GenBank mRNA | AB193481.1 | - | 1 | 235 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |