Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048028.1 | NP_001041493.1 | 3 - 4 | 333 | 79 | |
XM_005633300.3 | XP_005633357.1 | 3 - 4 | 459 | 79 | ||
XM_022407438.1 | XP_022263146.1 | 3 - 4 | 378 | 79 | ||
GenBank mRNA | AB210085.1 | - | 3 - 4 | 333 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons |