Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002939.1 | NP_001002939.1 | 12 - 13 | 1885 | 101 | |
XM_005622216.3 | XP_005622273.1 | 13 - 14 | 2368 | 101 | ||
XM_005622217.3 | XP_005622274.1 | 13 - 14 | 2306 | 101 | ||
GenBank mRNA | AF005360.1 | - | 12 - 13 | 1885 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |