Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002939.1 | NP_001002939.1 | 13 - 14 | 1961 | 97 | |
XM_005622216.3 | XP_005622273.1 | 14 - 15 | 2444 | 97 | ||
XM_005622217.3 | XP_005622274.1 | 14 - 15 | 2382 | 97 | ||
GenBank mRNA | AF005360.1 | - | 13 - 14 | 1961 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |