Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003339.1 | NP_001003339.1 | 6 - 7 | 969 | 70 | |
GenBank mRNA | AF029978.1 | - | 6 - 7 | 967 | 70 | |
AF029979.1 | - | 6 - 7 | 969 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |