Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLA-DQB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014381.1 | NP_001014403.1 | 3 - 4 | 663 | 79 | |
GenBank mRNA | AF043908.1 | - | 3 - 4 | 663 | 79 | |
M90802.1 | - | 2 - 3 | 528 | 79 | ||
M90803.1 | - | 2 - 3 | 528 | 79 | ||
M90804.1 | - | 2 - 3 | 528 | 79 | ||
M90805.1 | - | 2 - 3 | 528 | 79 | ||
S53615.1 | - | 2 - 3 | 528 | 79 | ||
Y07945.1 | - | 528 | 79 | |||
Y07946.1 | - | 528 | 79 |
Gene Symbol: LOC100856137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639415.4 | XP_003639463.1 | 3 - 4 | 752 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |