Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197044.1 | NP_001183973.1 | 5 - 6 | 835 | 115 | |
GenBank mRNA | AF049909.1 | - | 5 - 6 | 816 | 115 | |
HG965164.1 | - | 5 - 6 | 824 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |