Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002988.1 | NP_001002988.1 | 1 - 2 | 110 | 116 | |
XM_005627020.3 | XP_005627077.1 | 2 - 3 | 218 | 116 | ||
GenBank mRNA | AF160250.1 | - | 1 - 2 | 110 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons |