Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003114.1 | NP_001003114.1 | 989 | 71 | ||
GenBank mRNA | AF217203.1 | - | 989 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |