Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003133.1 | NP_001003133.1 | 9 - 10 | 1500 | 80 | |
GenBank mRNA | AF239824.1 | - | 9 - 10 | 1500 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |