Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V0E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003128.1 | NP_001003128.1 | 1 - 2 | 142 | 74 | |
GenBank mRNA | AF258614.1 | - | 1 - 2 | 142 | 74 | |
AF343440.1 | - | 1 - 2 | 132 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |