Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003084.1 | NP_001003084.1 | 5 - 6 | 888 | 66 | |
GenBank mRNA | AF325357.1 | - | 5 - 6 | 888 | 66 | |
AF390108.1 | - | 5 - 6 | 885 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |