Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003093.1 | NP_001003093.1 | 2 - 3 | 450 | 112 | |
XM_005625674.2 | XP_005625731.1 | 4 - 5 | 669 | 112 | ||
XM_022423980.1 | XP_022279688.1 | 4 - 5 | 669 | 112 | ||
GenBank mRNA | AF350327.1 | - | 2 - 3 | 450 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |