Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003022.1 | NP_001003022.1 | 2 - 3 | 4109 | 85 | |
GenBank mRNA | AF483595.1 | - | 2 - 3 | 4109 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |