Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003041.1 | NP_001003041.1 | 4 - 5 | 288 | 63 | |
GenBank mRNA | AF506750.1 | - | 4 - 5 | 288 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Inventoried Probe spans exons |