Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003032.1 | NP_001003032.1 | 3 - 4 | 336 | 76 | |
GenBank mRNA | AF525132.1 | - | 3 - 4 | 336 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |