Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003032.1 | NP_001003032.1 | 7 - 8 | 769 | 85 | |
GenBank mRNA | AF525132.1 | - | 7 - 8 | 769 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |