Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003029.1 | NP_001003029.1 | 2 - 3 | 429 | 75 | |
GenBank mRNA | AF525493.1 | - | 2 - 3 | 429 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |