Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLUL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002965.2 | NP_001002965.2 | 1041 | 92 | ||
NM_001293224.1 | NP_001280153.1 | 1 | 921 | 92 | ||
GenBank mRNA | AF544242.1 | - | 1 | 1045 | 92 | |
DN875727.1 | - | 1 | 283 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |