Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELSPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002931.1 | NP_001002931.1 | 2 - 3 | 152 | 70 | |
XM_022403243.1 | XP_022258951.1 | 2 - 3 | 265 | 70 | ||
XM_022404230.1 | XP_022259938.1 | 2 - 3 | 255 | 70 | ||
XM_022427808.1 | XP_022283516.1 | 2 - 3 | 251 | 70 | ||
GenBank mRNA | AJ278477.1 | - | 2 - 3 | 152 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |