Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287105.1 | NP_001274034.1 | 3 - 4 | 405 | 94 | |
NM_001287106.1 | NP_001274035.1 | 3 - 4 | 376 | 94 | ||
GenBank mRNA | AJ388514.1 | - | 1 - 2 | 92 | 94 | |
CO658560.1 | - | 2 - 3 | 205 | 94 | ||
DN863506.1 | - | 2 - 3 | 220 | 94 | ||
DN871196.1 | - | 3 - 4 | 376 | 94 | ||
GR889127.1 | - | 3 - 4 | 406 | 94 | ||
GR894797.1 | - | 3 - 4 | 354 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |