Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002953.1 | NP_001002953.1 | 2 - 3 | 281 | 55 | |
GenBank mRNA | AJ388551.2 | - | 2 - 3 | 281 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |