Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002954.1 | NP_001002954.1 | 2 - 3 | 239 | 63 | |
XM_005627964.3 | XP_005628021.1 | 2 - 3 | 209 | 63 | ||
XM_014118645.2 | XP_013974120.1 | 2 - 3 | 1095 | 63 | ||
XM_022426488.1 | XP_022282196.1 | 1 - 2 | 1314 | 63 | ||
XM_022426489.1 | XP_022282197.1 | 3 - 4 | 440 | 63 | ||
XM_022426490.1 | XP_022282198.1 | 3 - 4 | 370 | 63 | ||
GenBank mRNA | AJ388557.1 | - | 2 - 3 | 239 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |