Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB41L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002952.1 | NP_001002952.1 | 11 - 12 | 1007 | 124 | |
XM_005631761.2 | XP_005631818.1 | 11 - 12 | 1033 | 124 | ||
XM_014121360.2 | XP_013976835.1 | 11 - 12 | 1033 | 124 | ||
XM_014121361.2 | XP_013976836.1 | 11 - 12 | 1092 | 124 | ||
XM_014121362.2 | XP_013976837.1 | 11 - 12 | 1034 | 124 | ||
XM_014121364.2 | XP_013976839.1 | 11 - 12 | 1034 | 124 | ||
XM_014121366.2 | XP_013976841.1 | 11 - 12 | 1033 | 124 | ||
XM_022405860.1 | XP_022261568.1 | 11 - 12 | 1033 | 124 | ||
GenBank mRNA | AJ401272.1 | - | 11 - 12 | 1007 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |