Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022416190.1 | XP_022271898.1 | 3 | 1825 | 85 | |
XM_022416191.1 | XP_022271899.1 | 3 | 1838 | 85 | ||
XM_022416192.1 | XP_022271900.1 | 2 | 1764 | 85 | ||
GenBank mRNA | DQ195093.1 | - | 1 | 1003 | 85 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |