Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022416190.1 | XP_022271898.1 | 3 | 1501 | 142 | |
XM_022416191.1 | XP_022271899.1 | 3 | 1514 | 142 | ||
XM_022416192.1 | XP_022271900.1 | 2 | 1440 | 142 | ||
GenBank mRNA | DQ195093.1 | - | 1 | 679 | 142 | |
U67201.1 | - | 1 | 139 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Canis familiaris Inventoried Both primers and probe map within a single exon |