Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012647.1 | NP_001012665.1 | 380 | 68 | ||
XM_005629756.3 | XP_005629813.1 | 681 | 68 | |||
XM_005629757.3 | XP_005629814.1 | 771 | 68 | |||
GenBank mRNA | AJ879585.1 | - | 380 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Inventoried |