Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003064.2 | NP_001003064.2 | 6 - 7 | 979 | 79 | |
GenBank mRNA | AY062037.1 | - | 6 - 7 | 885 | 79 | |
DN399649.1 | - | 3 - 4 | 404 | 79 | ||
DN746677.1 | - | 257 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |