Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002962.1 | NP_001002962.1 | 1 - 2 | 196 | 146 | |
GenBank mRNA | AY251013.1 | - | 1 - 2 | 196 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |