Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002989.1 | NP_001002989.1 | 191 | 92 | ||
GenBank mRNA | AY266681.1 | - | 191 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris |