Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003386.2 | NP_001003386.2 | 6 - 7 | 1287 | 82 | |
GenBank mRNA | AY318943.1 | - | 6 - 7 | 1281 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |