Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002941.1 | NP_001002941.1 | 2 - 3 | 1069 | 80 | |
GenBank mRNA | AF129000.1 | - | 1 - 2 | 80 | ||
AY336053.1 | - | 2 - 3 | 1069 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |