Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286850.1 | NP_001273779.1 | 7 - 8 | 940 | 63 | |
NM_001286851.1 | NP_001273780.1 | 7 - 8 | 943 | 63 | ||
GenBank mRNA | AY375181.1 | - | 2 - 3 | 216 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |