Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002936.1 | NP_001002936.1 | 4 - 5 | 742 | 78 | |
XM_005631023.2 | XP_005631080.1 | 5 - 6 | 898 | 78 | ||
XM_022405056.1 | XP_022260764.1 | 5 - 6 | 1146 | 78 | ||
GenBank mRNA | AY533243.1 | - | 4 - 5 | 742 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |