Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629550.3 | XP_005629607.1 | 1559 | 97 | ||
XM_005629551.3 | XP_005629608.1 | 1727 | 97 | |||
GenBank mRNA | AY545218.2 | - | 1814 | 97 | ||
AY545219.2 | - | 1894 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris |