Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003383.1 | NP_001003383.1 | 3 - 4 | 250 | 88 | |
XM_005625680.3 | XP_005625737.1 | 3 - 4 | 311 | 88 | ||
XM_005625682.3 | XP_005625739.1 | 3 - 4 | 311 | 88 | ||
XM_005625683.3 | XP_005625740.1 | 3 - 4 | 311 | 88 | ||
XM_022423987.1 | XP_022279695.1 | 3 - 4 | 343 | 88 | ||
GenBank mRNA | AY553983.1 | - | 3 - 4 | 250 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |