Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003383.1 | NP_001003383.1 | 5 - 6 | 605 | 109 | |
XM_005625680.3 | XP_005625737.1 | 5 - 6 | 693 | 109 | ||
XM_005625682.3 | XP_005625739.1 | 5 - 6 | 666 | 109 | ||
GenBank mRNA | AY553983.1 | - | 5 - 6 | 605 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |