Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003194.2 | NP_001003194.1 | 1 - 2 | 99 | 62 | |
XM_005640752.2 | XP_005640809.1 | 1 - 2 | 177 | 62 | ||
GenBank mRNA | AY630442.1 | - | 1 - 2 | 99 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |