Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008552.1 | NP_001008552.1 | 386 | 113 | ||
GenBank mRNA | AY764285.1 | - | 386 | 113 | ||
GU563722.1 | - | 133 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |