Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011886.1 | NP_001011886.1 | 4 - 5 | 471 | 103 | |
GenBank mRNA | AY871313.1 | - | 4 - 5 | 471 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |