Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCER1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110766.2 | NP_001104236.2 | 2 - 3 | 80 | 60 | |
GenBank mRNA | D16413.1 | - | 2 - 3 | 88 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |