Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001018035.1 | NP_001018045.1 | 3 - 4 | 615 | 91 | |
XM_005628839.3 | XP_005628896.1 | 3 - 4 | 903 | 91 | ||
XM_005628841.3 | XP_005628898.1 | 2 - 3 | 673 | 91 | ||
XM_005628842.3 | XP_005628899.1 | 2 - 3 | 436 | 91 | ||
XM_014119231.2 | XP_013974706.1 | 3 - 4 | 824 | 91 | ||
XM_022427665.1 | XP_022283373.1 | 3 - 4 | 614 | 91 | ||
XM_022427666.1 | XP_022283374.1 | 3 - 4 | 611 | 91 | ||
XM_022427667.1 | XP_022283375.1 | 2 - 3 | 610 | 91 | ||
XM_022427668.1 | XP_022283376.1 | 2 - 3 | 449 | 91 | ||
GenBank mRNA | DQ008575.1 | - | 3 - 4 | 615 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |