Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR3C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014119440.2 | XP_013974915.1 | 8 - 9 | 3061 | 62 | |
XM_014119441.2 | XP_013974916.1 | 8 - 9 | 2813 | 62 | ||
XM_022403487.1 | XP_022259195.1 | 7 - 8 | 1246 | 62 | ||
GenBank mRNA | DQ195095.1 | - | 4 - 5 | 532 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |