Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003341.1 | NP_001003341.1 | 878 | 89 | ||
GenBank mRNA | DQ202370.1 | - | 520 | 89 | ||
U49457.1 | - | 878 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris |